6-Phenyl-m-anisidine hydrochloride

Supplier: Matrix Scientific

056804-1G
101911-434EA 207.69 USD
101911-434
6-Phenyl-m-anisidine hydrochloride
6-Phenyl-m-anisidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₄ClNO
Melting Pt: 196…198 °C
MDL Number: MFCD00270121
CAS Number: 107624-16-8

Order Now

Specification Test Results

Melting point 196-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR