4-(4-Phenoxyphenyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056798-1G
101911-422EA 76.92 USD
101911-422
4-(4-Phenoxyphenyl)-3-thiosemicarbazide
4-(4-Phenoxyphenyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₁₃H₁₃N₃OS
Melting Pt: 140 °C (dec.)
MDL Number: MFCD00041299
CAS Number: 206761-85-5

Order Now

Specification Test Results

Melting point 140°C (dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR