4-Phenoxybenzhydrazide

Supplier: Matrix Scientific

056797-5G
101911-420EA 112.7 USD
101911-420
4-Phenoxybenzhydrazide
4-Phenoxybenzhydrazide

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₂
Melting Pt: 125…127 °C
MDL Number: MFCD00153894
CAS Number: 143667-36-1

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR