2-Pentyloxy-5-phenylaniline

Supplier: Matrix Scientific

056793-1G
101911-412EA 56.92 USD
101911-412
2-Pentyloxy-5-phenylaniline
2-Pentyloxy-5-phenylaniline

Hazard: Irritant

Formula: C₁₇H₂₁NO
Melting Pt: 47…49 °C
MDL Number: MFCD00270119
CAS Number: 206761-83-3

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR