1-Pentafluorophenyl-2-thiourea

Supplier: Matrix Scientific

056791-1G
101911-408EA 153.85 USD
101911-408
1-Pentafluorophenyl-2-thiourea
1-Pentafluorophenyl-2-thiourea

Hazard: Irritant

Formula: C₇H₃F₅N₂S
Melting Pt: 156…158 °C
MDL Number: MFCD00041149
CAS Number: 715-60-6

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR