4-(Octyl)thiosemicarbazide
Supplier: Matrix Scientific
056789-1G
101911-404EA
64.62
USD
101911-404
4-(Octyl)thiosemicarbazide
4-(Octyl)thiosemicarbazide
Hazard: Irritant
Formula:
C₉H₂₁N₃S Melting Pt: 47…49 °C |
MDL Number:
MFCD00041382 CAS Number: 13207-36-8 |
Specification Test Results
Melting point | 47-49°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...