4-(1-Naphthyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056779-1G
101911-384EA 103.08 USD
101911-384
4-(1-Naphthyl)-3-thiosemicarbazide
4-(1-Naphthyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₁₁H₁₁N₃S
Melting Pt: 139…141 °C (dec.)
MDL Number: MFCD00060343
CAS Number: 42135-78-4

Order Now

Specification Test Results

Melting point 139-141°C (

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR