4-(3-Morpholinopropyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056776-1G
101911-378EA 49.23 USD
101911-378
4-(3-Morpholinopropyl)-3-thiosemicarbazide
4-(3-Morpholinopropyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₈H₁₈N₄OS
Melting Pt: 135…137 °C
MDL Number: MFCD00023400
CAS Number: 32813-48-2

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR