1-[2-(Methylthio)phenyl]-2-thiourea

Supplier: Matrix Scientific

056773-1G
101911-372EA 153.85 USD
101911-372
1-[2-(Methylthio)phenyl]-2-thiourea
1-[2-(Methylthio)phenyl]-2-thiourea

Hazard: Irritant

Formula: C₈H₁₀N₂S₂
Melting Pt: 168…170 °C
MDL Number: MFCD00060434
CAS Number: 59084-10-5

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR