4-(3,4-Methylenedioxybenzyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056762-1G
101911-350EA 106.15 USD
101911-350
4-(3,4-Methylenedioxybenzyl)-3-thiosemicarbazide
4-(3,4-Methylenedioxybenzyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₉H₁₁N₃O₂S
Melting Pt: 158…160 °C
MDL Number: MFCD00060506
CAS Number: 206761-70-8

Order Now

Specification Test Results

Melting point 158-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR