Methyl-4-phenylbenzoate ≥99%

Supplier: Matrix Scientific

Synonyms: p-Phenylbenzoic acid-OMeMethyl [1,1'-biphenyl]-4-carboxylateMethyl biphenyl-4-carboxylate

056760-10G
101911-344EA 150.77 USD
101911-344
Methyl-4-phenylbenzoate ≥99%
Methyl 4-phenylbenzoate

Hazard: Irritant

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD00017200
CAS Number: 720-75-2

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR