1-(4-Methylbenzyl)-2-thiourea

Supplier: Matrix Scientific

056759-1G
101911-342EA 53.85 USD
101911-342
1-(4-Methylbenzyl)-2-thiourea
1-(4-Methylbenzyl)-2-thiourea

Hazard: Irritant

Formula: C₉H₁₂N₂S
Melting Pt: 120…122 °C
MDL Number: MFCD00060468
CAS Number: 103854-74-6

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR