4-(3-Methoxypropyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056758-1G
101911-340EA 60 USD
101911-340
4-(3-Methoxypropyl)-3-thiosemicarbazide
4-(3-Methoxypropyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₅H₁₃N₃OS
Melting Pt: 37…40 °C
MDL Number: MFCD00060598
CAS Number: 71058-32-7

Order Now

Specification Test Results

Melting point 37-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR