4-(4-Methoxybenzyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056748-1G
101911-320EA 98.46 USD
101911-320
4-(4-Methoxybenzyl)-3-thiosemicarbazide
4-(4-Methoxybenzyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₉H₁₃N₃OS
Melting Pt: 131…133 °C
MDL Number: MFCD00060591
CAS Number: 16735-76-5

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR