4-(2-Furfuryl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056733-1G
101911-290EA 98.46 USD
101911-290
4-(2-Furfuryl)-3-thiosemicarbazide
4-(2-Furfuryl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₆H₉N₃OS
Melting Pt: 119…121 °C
MDL Number: MFCD00060333
CAS Number: 96860-19-4

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR