1-(4-Ethoxycarbonylphenyl)-2-thiourea
Supplier: Matrix Scientific
056726-5G
101911-278EA
312.31
USD
101911-278
1-(4-Ethoxycarbonylphenyl)-2-thiourea
1-(4-Ethoxycarbonylphenyl)-2-thiourea
Hazard: Irritant
Formula:
C₁₀H₁₂N₂O₂S Melting Pt: 157…159 °C |
MDL Number:
MFCD00060458 CAS Number: 23051-16-3 |
Specification Test Results
Melting point | 157-159°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...