1-(3-Ethoxycarbonylphenyl)-2-thiourea

Supplier: Matrix Scientific

056725-1G
101911-276EA 95.38 USD
101911-276
1-(3-Ethoxycarbonylphenyl)-2-thiourea
1-(3-Ethoxycarbonylphenyl)-2-thiourea

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂S
Melting Pt: 129…131 °C
MDL Number: MFCD00060449
CAS Number: 20967-87-7

Order Now

Specification Test Results

Melting point 129-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR