1-Dodecyl-3-phenyl-2-thiourea

Supplier: Matrix Scientific

056722-5G
101911-270EA 129.23 USD
101911-270
1-Dodecyl-3-phenyl-2-thiourea
1-Dodecyl-3-phenyl-2-thiourea

Hazard: Irritant

Formula: C₁₉H₃₂N₂S
Melting Pt: 68…71 °C
MDL Number: MFCD00022121
CAS Number: 63980-78-9

Order Now

Specification Test Results

Melting point 68-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR