3,4-Dimethoxybenzyl isothiocyanate

Supplier: Matrix Scientific

056711-1G
101911-248EA 107.69 USD
101911-248
3,4-Dimethoxybenzyl isothiocyanate
3,4-Dimethoxybenzyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₁₀H₁₁NO₂S
Melting Pt: 29…32 °C
MDL Number: MFCD00041113
CAS Number: 14596-50-0

Order Now

Specification Test Results

Melting point 29-32°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR