Cinnamoyl isothiocyanate

Supplier: Matrix Scientific

056691-5G
101911-212EA 221.54 USD
101911-212
Cinnamoyl isothiocyanate
Cinnamoyl isothiocyanate

Hazard: Irritant-Harmful/Lachrymator

Formula: C₁₀H₇NOS
Melting Pt: 43…45 °C
MDL Number: MFCD00046813
CAS Number: 19495-08-0

Order Now

Specification Test Results

Melting point 43-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR