(-)-4'-Chlorotartranilic acid

Supplier: Matrix Scientific

Synonyms: (2S,3S)-4-((4-Chlorophenyl)amino)-2,3-dihydroxy-4-oxobutanoic acid

056690-1G
101911-210EA 121.54 USD
101911-210
(-)-4'-Chlorotartranilic acid
(-)-4'-Chlorotartranilic acid

Hazard: Irritant

Formula: C₁₀H₁₀ClNO₅
MW: 259.65 g/mol
Melting Pt: 193…195 °C (dec.)
MDL Number: MFCD00070634
CAS Number: 46834-56-4

Order Now

Specification Test Results

Melting point 193-195°C (

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR