1-(5-Chloro-2-methylphenyl)-2-thiourea

Supplier: Matrix Scientific

056682-10G
101911-194EA 169.23 USD
101911-194
1-(5-Chloro-2-methylphenyl)-2-thiourea
1-(5-Chloro-2-methylphenyl)-2-thiourea

Hazard: Irritant

Formula: C₈H₉ClN₂S
Melting Pt: 160…161 °C
MDL Number: MFCD00060444
CAS Number: 72806-61-2

Order Now

Specification Test Results

Melting point 160-161°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR