5-Chloro-2,4-dimethoxyphenyl isothiocyanate

Supplier: Matrix Scientific

056676-1G
101911-182EA 149.23 USD
101911-182
5-Chloro-2,4-dimethoxyphenyl isothiocyanate
5-Chloro-2,4-dimethoxyphenyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₉H₈ClNO₂S
Melting Pt: 92…94 °C
MDL Number: MFCD00060373
CAS Number: 40046-27-3

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR