1-(4-Carboxyphenyl)-2-thiourea

Supplier: Matrix Scientific

Synonyms: 4-Thioureidobenzoic acid

056673-5G
101911-176EA 169.23 USD
101911-176
1-(4-Carboxyphenyl)-2-thiourea
1-(4-Carboxyphenyl)-2-thiourea

Hazard: Irritant

Formula: C₈H₈N₂O₂S
MW: 196.23 g/mol
Melting Pt: 255 °C
MDL Number: MFCD00041187
CAS Number: 7366-56-5

Order Now

Specification Test Results

Melting point 255°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR