4-(4-Carboxyphenyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056671-1G
101911-172EA 123.08 USD
101911-172
4-(4-Carboxyphenyl)-3-thiosemicarbazide
4-(4-Carboxyphenyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₈H₉N₃O₂S
Melting Pt: 204…206 °C
MDL Number: MFCD00060584
CAS Number: 206559-48-0

Order Now

Specification Test Results

Melting point 204-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR