5-Bromo-2-furoic acid hydrazide

Supplier: Matrix Scientific

Synonyms: 5-Bromofuran-2-carbohydrazide

056662-5G
101911-156EA 135.38 USD
101911-156
5-Bromo-2-furoic acid hydrazide
5-Bromo-2-furoic acid hydrazide

Hazard: Irritant

Formula: C₅H₅BrN₂O₂
MW: 205.02 g/mol
Melting Pt: 136…138 °C
MDL Number: MFCD01310787
CAS Number: 89282-37-1

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR