4-Bromo-2,6-dimethylphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056660-1G
101911-152EA 47.15 USD
101911-152
4-Bromo-2,6-dimethylphenyl isothiocyanate ≥98%
4-Bromo-2,6-dimethylphenyl isothiocyanate

Hazard: Irritant-Harmful

Formula: C₉H₈BrNS
Melting Pt: 77…80 °C
MDL Number: MFCD00041088
CAS Number: 32265-82-0

Order Now

Specification Test Results

Melting point 77-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR