4-Bromo-3-chlorobenzhydrazide

Supplier: Matrix Scientific

056658-1G
101911-148EA 198.46 USD
101911-148
4-Bromo-3-chlorobenzhydrazide
4-Bromo-3-chlorobenzhydrazide

Hazard: Irritant

Formula: C₇H₆BrClN₂O
Melting Pt: 171…173 °C
MDL Number: MFCD00270096
CAS Number: 148993-19-5

Order Now

Specification Test Results

Melting point 171-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR