4-Bromo-2-chlorobenzhydrazide

Supplier: Matrix Scientific

056657-1G
101911-146EA 151.8 USD
101911-146
4-Bromo-2-chlorobenzhydrazide
4-Bromo-2-chlorobenzhydrazide

Hazard: Irritant

Formula: C₇H₆BrClN₂O
Melting Pt: 173…175 °C
MDL Number: MFCD00270095
CAS Number: 206559-39-9

Order Now

Specification Test Results

Melting point 173-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR