2,6-Bis(thiocyanatomethyl)-4-methylanisole

Supplier: Matrix Scientific

056655-1G
101911-142EA 118.46 USD
101911-142
2,6-Bis(thiocyanatomethyl)-4-methylanisole
2,6-Bis(thiocyanatomethyl)-4-methylanisole

Hazard: Irritant

Formula: C₁₂H₁₂N₂OS₂
Melting Pt: 122…124 °C
CAS Number: 206559-38-8

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR