4-(4-Benzyloxyphenyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056648-1G
101911-128EA 120 USD
101911-128
4-(4-Benzyloxyphenyl)-3-thiosemicarbazide
4-(4-Benzyloxyphenyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₁₄H₁₅N₃OS
Melting Pt: 165…168 °C (dec.)
MDL Number: MFCD00041300
CAS Number: 206559-37-7

Order Now

Specification Test Results

Melting point 165-168°C (

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR