3-Benzyloxybenzhydrazide

Supplier: Matrix Scientific

Synonyms: 3-(Benzyloxy)benzohydrazide

056647-5G
101911-126EA 212.31 USD
101911-126
3-Benzyloxybenzhydrazide
3-Benzyloxybenzhydrazide

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₂
MW: 242.2732 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD01310784
CAS Number: 228419-13-4

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR