2-Amino-N-cyclohexylbenzamide

Supplier: Matrix Scientific

056637-5G
101911-104EA 138.46 USD
101911-104
2-Amino-N-cyclohexylbenzamide
2-Amino-N-cyclohexylbenzamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
Melting Pt: 153…156 °C
MDL Number: MFCD00060339
CAS Number: 56814-11-0

Order Now

Specification Test Results

Melting point 153-156°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR