2-Phenoxybenzhydrazide ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Phenoxybenzohydrazide

056626-5G
101911-082EA 157.55 USD
101911-082
2-Phenoxybenzhydrazide ≥98%
2-Phenoxybenzhydrazide

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₂
MW: 228.25 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD00017048
CAS Number: 43038-37-5

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR