4-Phenylazophenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

056616-5G
101911-052EA 181.54 USD
101911-052
4-Phenylazophenyl isothiocyanate ≥98%
4-Phenylazophenyl isothiocyanate

Hazard: Irritant-Harmful/Lachrymator

Formula: C₁₃H₉N₃S
Melting Pt: 94…96 °C
MDL Number: MFCD00041092
CAS Number: 7612-96-6

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR