4-(2,4,6-Trifluorophenyl)-3-thiosemicarbazide

Supplier: Matrix Scientific

056589-1G
101910-970EA 144.62 USD
101910-970
4-(2,4,6-Trifluorophenyl)-3-thiosemicarbazide
4-(2,4,6-Trifluorophenyl)-3-thiosemicarbazide

Hazard: Irritant

Formula: C₇H₆F₃N₃S
Melting Pt: 204…205 °C
MDL Number: MFCD00060564
CAS Number: 206761-92-4

Order Now

Specification Test Results

Melting point 204-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR