1-(2-(Trifluoromethyl)phenyl)thiourea
Supplier: Matrix Scientific
Synonyms:
N-[2-(Trifluoromethyl)phenyl]thiourea
056588-5G
101910-968EA
55.38
USD
101910-968
1-(2-(Trifluoromethyl)phenyl)thiourea
1-(2-(Trifluoromethyl)phenyl)thiourea
Hazard: Irritant
Formula:
C₈H₇F₃N₂S MW: 220.22 Melting Pt: 158…159 °C |
MDL Number:
MFCD00041161 CAS Number: 1736-71-6 |
Specification Test Results
Melting point | 158-159°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...