4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide

Supplier: Matrix Scientific

056585-1G
101910-958EA 56.92 USD
101910-958
4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide
4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide

Hazard: Irritant

Formula: C₈H₈F₃N₃S
Melting Pt: 120…122 °C
MDL Number: MFCD00041291
CAS Number: 20069-30-1

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR