3-(Benzyloxy)phenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Benzyloxyphenylisothiocyanate

056548-1G
101910-850EA 80 USD
101910-850
3-(Benzyloxy)phenyl isothiocyanate ≥98%
3-(Benzyloxy)phenyl isothiocyanate

Hazard: Toxic/Corrosive/Moisture Sensitive

Formula: C₁₄H₁₁NOS
MW: 241.31 g/mol
Melting Pt: 49…51 °C
MDL Number: MFCD00060374
CAS Number: 206559-36-6

Order Now

Specification Test Results

Melting point 49-51°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR