3,4-(Methylenedioxy)phenylacetonitrile ≥98%

Supplier: Matrix Scientific

056547-5G
101910-848EA 61.54 USD
101910-848
3,4-(Methylenedioxy)phenylacetonitrile ≥98%
3,4-(Methylenedioxy)phenylacetonitrile

Hazard: Irritant

Formula: C₉H₇NO₂
Boiling Pt: 160 °C (10 Torr)
Melting Pt: 49 °C
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 43-45°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR