4-Phenylmorpholin-3-one ≥95%

Supplier: Matrix Scientific

056506-5G 056506-1G
101910-776EA 369.23 USD
101910-776 101910-774
4-Phenylmorpholin-3-one ≥95%
4-Phenylmorpholin-3-one

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
Melting Pt: 114…116 °C
MDL Number: MFCD00624822
CAS Number: 29518-11-4

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR