Methyl-1,3-dimethyl-2-oxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylate ≥95%

Supplier: Matrix Scientific

056500-500MG 056500-1G
101910-754EA 1163.08 USD
101910-754 101910-752
Methyl-1,3-dimethyl-2-oxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylate ≥95%
Methyl-1,3-dimethyl-2-oxo-1,2,3,4-tetrahydroquinoxaline-6-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₃
Melting Pt: 168…170 °C
MDL Number: MFCD16890152
CAS Number: 1246738-25-9

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR