4-Chloro-2-(pyrimidin-2-yl)-6-(trifluoromethyl)pyrimidine ≥95%
Supplier: Matrix Scientific
056472-500MG
056472-1G
101910-634EA
698.46
USD
101910-634
101910-632
4-Chloro-2-(pyrimidin-2-yl)-6-(trifluoromethyl)pyrimidine ≥95%
4-Chloro-2-(pyrimidin-2-yl)-6-(trifluoromethyl)pyrimidine
Hazard: Irritant
Formula:
C₉H₄ClF₃N₄ Melting Pt: 88…89 °C |
MDL Number:
MFCD16890136 CAS Number: 438249-97-9 |
Specification Test Results
Melting point | 88-89°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...