6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%

056470-1G 056470-500MG
101910-626EA 602.6 USD
101910-626 101910-624
6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
6-Bromo-2-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile

Hazard: Irritant

Formula: C₈H₅BrN₄
Melting Pt: 223…225 °C
MDL Number: MFCD16890140
CAS Number: 1242268-30-9

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR