tert-Butyl-3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
056455-500MG
056455-1G
101910-568EA
232.31
USD
101910-568
101910-566
tert-Butyl-3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥95%
tert-Butyl-3-formyl-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₃ MW: 246.27 g/mol Melting Pt: 125…127 °C |
MDL Number:
MFCD11045434 CAS Number: 144657-66-9 |
Specification Test Results
Melting point | 125-127°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...