tert-Butyl-4-(6-chloropyridin-3-yl)piperazine-1-carboxylate ≥95%
Supplier: Matrix Scientific
056453-1G
056453-500MG
101910-558EA
384.62
USD
101910-558
101910-560
tert-Butyl-4-(6-chloropyridin-3-yl)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(6-chloropyridin-3-yl)piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₂₀ClN₃O₂ MW: 297.79 g/mol Melting Pt: 118…119 °C |
MDL Number:
MFCD12923074 CAS Number: 633283-53-1 |
Specification Test Results
Melting point | 118-119°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...