1-(tert-Butoxycarbonyl)-2-(chloromethyl)benzimidazole ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-(chloromethyl)-1H-benzo[d]imidazole-1-carboxylate

056448-500MG
101910-542EA 353.85 USD
101910-542
1-(tert-Butoxycarbonyl)-2-(chloromethyl)benzimidazole ≥97%
1-(tert-Butoxycarbonyl)-2-(chloromethyl)benzimidazole

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂O₂
MW: 266.73 g/mol
Melting Pt: 80…82 °C
MDL Number: MFCD10697638
CAS Number: 163798-87-6

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR