1-Methyl-2-phenyl-1H-imidazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

056434-500MG
101910-488EA 1109.23 USD
101910-488
1-Methyl-2-phenyl-1H-imidazole-5-carbaldehyde ≥95%
1-Methyl-2-phenyl-1H-imidazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₁₀N₂O
Melting Pt: 94…96 °C
MDL Number: MFCD08703273
CAS Number: 94938-03-1

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR