Methyl-1-ethyl-1H-1,3-benzodiazole-6-carboxylate ≥95%

Supplier: Matrix Scientific

056430-500MG
101910-476EA 556.92 USD
101910-476
Methyl-1-ethyl-1H-1,3-benzodiazole-6-carboxylate ≥95%
Methyl-1-ethyl-1H-1,3-benzodiazole-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₂
Melting Pt: 85…87 °C
MDL Number: MFCD13195704
CAS Number: 1199773-11-9

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR