Methyl-2-[4-benzyl-2-(propan-2-yl)piperazin-1-yl]-5-nitrobenzoate ≥95%

Supplier: Matrix Scientific

056424-500MG
101910-454EA 921.54 USD
101910-454
Methyl-2-[4-benzyl-2-(propan-2-yl)piperazin-1-yl]-5-nitrobenzoate ≥95%
Methyl-2-[4-benzyl-2-(propan-2-yl)piperazin-1-yl]-5-nitrobenzoate

Hazard: Irritant

Formula: C₂₂H₂₇N₃O₄
Melting Pt: 96…98 °C
MDL Number: MFCD16710280
CAS Number: 1242268-07-0

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR